Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEX19 Rabbit anti-Human, Rat, Clone: 9O7Q9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PEX19 |
---|---|
Clone | 9O7Q9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PEX19 Monoclonal antibody specifically detects PEX19 in Human, Rat samples. It is validated for Western BlotSpecifications
PEX19 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5824 | |
IgG | |
Affinity purified |
9O7Q9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
D1S2223Ehousekeeping gene, 33kD, HK3333 kDa housekeeping protein, Peroxin-19, peroxisomal biogenesis factor 19, Peroxisomal farnesylated proteinFLJ55296, PMP1, PMPI, PXFperoxin-19, PXMP1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PEX19 (P40855). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title