Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX2 Polyclonal Antibody

Catalog No. PIPA5121691
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121691 20 μL
PIPA5119334 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121691 Supplier Invitrogen™ Supplier No. PA5121691
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PEX2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PEX2
Gene Accession No. P24392, P28328
Gene Alias 35 kDa peroxisomal membrane protein; D3Ertd138e; Paf1; PAF-1; PBD5A; PBD5B; peroxin 2; Peroxin-2; peroxisomal biogenesis factor 2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; peroxisome assembly factor 1; peroxisome assembly factor-1; peroxisome biogenesis factor 2; Pex2; PMP3; Pmp35; Pxmp3; RING finger protein 72; RNF72; Zellweger syndrome homolog; ZWS3
Gene Symbols PEX2
Host Species Rabbit
Immunogen Recombinant protein Peroxisomal Biogenesis Factor 2. The antigen corresponds to amino acid range 233-305 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29534, 5828
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.