Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584362
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584362 Supplier Invitrogen™ Supplier No. PA584362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLLDLEQKLK EIRNLVEQHK SSSWINKDGS KPLLCHYMMP DEETPLAVQA CGLSPRDITT IKLLNETRDM LESPDFSTVL N Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 84%.

Peroxin 3, also known as PEX3 (peroxisomal biogenesis factor 3) or TRG18, is a 373 amino acid multi-pass membrane protein that localizes to peroxisomes and belongs to the Peroxin family. Expressed ubiquitously, Peroxin 3 interacts with Peroxin 19 and is involved in peroxisome biosynthesis and membrane vesicle assembly, as well as in the maintenance of peroxisomal integrity. Additionally, Peroxin 3 acts as a docking factor for Peroxin 19 and is required for the import of peroxisomal proteins. Defects in the gene encoding Peroxin 3 are the cause of peroxisome biogenesis disorder complementation group 12 (PBD-CG12) and Zellweger syndrome (ZwS), both of which arise from a failure of peroxisomal protein import.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PEX3
Gene Accession No. P56589
Gene Alias 1700014F15Rik; 2810027F19Rik; 2900010N04Rik; PBD10A; Peroxin3; peroxin-3; peroxisomal assembly protein 3; peroxisomal assembly protein PEX3; Peroxisomal biogenesis factor 3; pex3; transformation-related protein 18; TRG18; zgc:56313
Gene Symbols PEX3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PEX3. Recombinant protein control fragment (Product #RP-103314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.