Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEX5 Monoclonal Antibody (OTI6C4), TrueMAB™, OriGene
SDP

Catalog No. 502542324
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2324 30 μL
50-167-2532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2324 Supplier OriGene Supplier No. TA501390S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PEX5 Monoclonal antibody specifically detects PEX5 in Canine, Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PEX5
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6C4
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Pex5
Gene Accession No. P50542
Gene Alias AW212715; ESTM1; LOW QUALITY PROTEIN: peroxisomal biogenesis factor 5; PBD2A; PBD2B; peroxin 5; peroxin-5; peroxisomal biogenesis factor 5; peroxisomal C-terminal targeting signal import receptor; peroxisomal targeting signal 1 (SKL type) receptor; peroxisomal targeting signal 1 receptor; peroxisomal targeting signal 1 receptor; peroxisomal biogenesis factor 5; peroxisomal targeting signal import receptor; peroxisomal targeting signal receptor 1; peroxisome biogenesis factor 5; peroxisome receptor 1; PEX5; Pex5_predicted; PTS1 BP; PTS1 receptor; PTS1-BP; PTS1R; Pxr1; PXR1P; RCDP5; X83306
Host Species Mouse
Immunogen Full length human recombinant protein of human PEX5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 486710, 5830
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.