Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578550 Supplier Invitrogen™ Supplier No. PA578550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted Reactivity: Human (93%), Rhesus Monkey (94%), Bovine (85%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Pex5l is an accessory subunit of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels, regulating their cell-surface expression and cyclic nucleotide dependence. Research studies have shown that Pex5l is expressed in the brain, testis and pancreas. Diseases associated with PEX5L include Rhizomelic Chondrodysplasia Punctata, Type 5 and Rhizomelic Chondrodysplasia Punctata, Type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX5L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene PEX5L
Gene Accession No. Q8C437, Q925N3
Gene Alias 1700016J08Rik; HCN channel auxiliary subunit; peroxin 2; peroxin-5-related protein; peroxisomal biogenesis factor 5 like; peroxisomal biogenesis factor 5-like; peroxisomal targeting signal 1 receptor-like; peroxisome biogenesis factor 5-like; Pex2; PEX2-related protein; Pex5l; PEX5-like protein; Pex5p-related protein; Pex5r; PEX5-related protein; PEX5RP; PXR2; PXR2B; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein; Trip8b; Trjip8b
Gene Symbols PEX5L
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of mouse PEX5L. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286937, 58869
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.