Learn More
Invitrogen™ PEX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587917
Description
Immunogen sequence: LLHGPPGTGK TLLAKAVATE CSLTFLSVKG PELINMYVGQ SEENVREVFA RARAAAPCII FFDELDSLAP SRGRSGDSGG VMDRVVSQLL AELDGLHSTQ DVFVIGATNR PDLLDPALLR PGRFDKLVFV GANEDRASQL RVLSAITRKF KLEPSVSLVN VLDCCPPQLT GADLYSLCSD AMTAALKRRV HDLEEGLEPG SSALMLTMED LLQAAARLQP SVSEQELLRY KRIQRKFAAC; Positive Samples: BT-474, K-562, Mouse liver, Mouse brain; Cellular Location: Cytoplasm, Peroxisome membrane.
PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
Specifications
PEX6 | |
Polyclonal | |
Unconjugated | |
Pex6 | |
AI132582; D130055I09Rik; HMLR2; mKIAA4177; PAF 2; Paf2; Paf-2; PBD4A; PDB4B; Peroxin 6; Peroxin-6; peroxisomal AAA-type ATPase 1; peroxisomal biogenesis factor 6; Peroxisomal-type ATPase 1; peroxisome assembly factor 2; peroxisome biogenesis factor 6; PEX6; PXAAA1; RP1-125M3.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
224824, 5190 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
4.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13608, Q99LC9 | |
Pex6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 741-980 of human PEX6 (NP_0002783). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.