Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522058 Supplier Invitrogen™ Supplier No. PA522058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HepG2. Predicted reactivity: Mouse (81%), Rat (80%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Pex6
Gene Accession No. Q13608
Gene Alias AI132582; D130055I09Rik; HMLR2; mKIAA4177; PAF 2; Paf2; Paf-2; PBD4A; PDB4B; Peroxin 6; Peroxin-6; peroxisomal AAA-type ATPase 1; peroxisomal biogenesis factor 6; Peroxisomal-type ATPase 1; peroxisome assembly factor 2; peroxisome biogenesis factor 6; PEX6; PXAAA1; RP1-125M3.3
Gene Symbols Pex6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PEX6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5190
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.