Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pexacerfont
GREENER_CHOICESDP

Catalog No. 645150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-515-0 50 mg
64-511-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 64-515-0 Supplier Tocris Bioscience™ Supplier No. 6451/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CRF1 antagonist; anxiolytic

Pexacerfont is a potent and selective corticotropin-releasing factor 1 receptor (CRF1) antagonist (IC50 = 6.1 nM). Displays >150 fold selectivity for CRF1 over CRF2b. Inhibits CRF-mediated adrenocorticotropic hormone (ACTH) release from pituitary cell culture (IC50 = 129 nM). Displays anxiolytic effects in rat behavioral paradigms.

Specifications

Chemical Name or Material 8-(6-Methoxy-2-methyl-3-pyridinyl)-2,7-dimethyl-N-[(1R)-1-methylpropyl]pyrazolo[1,5-a]-1,3,5-triazin-4-amine
CAS 459856-18-9
Quantity 50 mg
Molecular Formula C18H24N6O
Formula Weight 340.43
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.