Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PF 04418948
GREENER_CHOICESDP

Catalog No. 481850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-185-0 50 mg
48-181-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-185-0 Supplier R&D Systems™ Supplier No. 4818/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective EP2 receptor antagonist

Potent EP2 receptor antagonist (IC50 = 16 nM for human EP2 receptors). Displays over 2000-fold selectivity for EP2 receptors over EP1, EP3, EP4, DP1 amd CRTH2 receptors; exhibits <30% binding at a diverse panel of GPCRs and ion channels at a concentration of 10 μM. Inhibits PGE2-induced increases in intracellular cAMP; reverses PGE2-invoked relaxation of mouse trachea (IC50 = 2.7 nM).

Chemical Identifiers

CAS 1078166-57-0
Molecular Formula C23H20FNO5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 409.41
Purity 98%
Molecular Weight (g/mol) 409.413
Chemical Name or Material PF 04418948
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.