Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 04620110
GREENER_CHOICE

Catalog No. 586325
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-632-5 25 mg
58-635 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 58-632-5 Supplier Tocris Bioscience™ Supplier No. 5863/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective DGAT-1 inhibitor; orally bioavailable

PF 04620110 is a potent and selective DGAT-1 inhibitor (IC50 = 19 nM). Exhibits >1000-fold selectivity for DGAT-1 over DGAT-2, and no significant activity against a range of other enzymes, ion channels and receptors. Reduces plasma triglyceride levels in a rat lipid challenge model. Orally bioavailable.

Specifications

Chemical Name or Material trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]cyclohexaneacetic acid
CAS 1109276-89-2
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C21H24N4O4
Formula Weight Observed MW: 396.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.