Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PF 04885614
GREENER_CHOICESDP

Catalog No. 491610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-161-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 49-161-0 Supplier R&D Systems™ Supplier No. 4916/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NaV1.8 channel blocker; active in vivo

Potent NaV1.8 inhibitor (IC50 = 53 nM for human NaV1.8 channel). Exhibits selectivity for hNaV1.8 over hNaV1.6, hNaV1.7, hNaV1.1, hNaV1.2 and hNaV1.5 (IC50 values are 4.2, 7.0, 11, 16 and 27 μM respectively). Orally bioavailable.

Chemical Identifiers

CAS 1480833-70-2
Molecular Formula C13H14F3N3O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 285.26
Purity >98%
Molecular Weight (g/mol) 285.27
Chemical Name or Material PF 04885614
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.