Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PF 06260933 dihydrochloride
GREENER_CHOICESDP

Catalog No. 575210 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-521-0 10 mg
57-525-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-521-0 Supplier R&D Systems™ Supplier No. 5752/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MAP4K4 (HGK) inhibitor; also inhibits MINK and TNIK

PF 06260933 dihydrochloride is a MAP4K4 (HGK) inhibitor (IC50 = 140 nM). Also inhibits MINK and TNIK (IC50 values are 8 and 13 nM, respectively). Improves fasting hyperglycemia in mice. Orally active.

Specifications

Chemical Name or Material 5-(4-Chlorophenyl)-[3,3 '-bipyridine]-6,6 '-diamine dihydrochloride
CAS 1883548-86-4
Synonym EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAP4K4, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, MEKKK4, mitogen-activa
Quantity 10 mg
Product Line Tocris
Target HGK/MAP4K4
Molecular Formula C16H13ClN40.2 HCl
Formula Weight 369.68
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.