Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 06281355, >98%

Catalog No. 600410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-041-0 10 mg
60-045-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-041-0 Supplier Tocris Bioscience™ Supplier No. 6004/10
This item is not returnable. View return policy

Selective myeloperoxidase (MPO) inhibitor

Selective myeloperoxidase (MPO) inhibitor (IC50 = 1.5 μM in LPS-stimulated human whole blood). Selective for MPO over a panel of 50 receptors, enzymes, ion channels and transporters, including thyroid peroxidase. Inhibits MPO activity in peritoneal lavage and plasma in a murine peritonitis model. Also reduces lung edema in a murine pulmonary immune complex vasculitis model. Orally bioavailable.

Chemical Identifiers

CAS 1435467-38-1
Molecular Formula C14H15N3O4S

Specifications

Quantity 10 mg
Solubility Soluble to 20mM in DMSO with gentle warming
Formula Weight 321.35
Purity >98%
Molecular Weight (g/mol) 321.351
Chemical Name or Material 6-(2,5-Dimethoxyphenyl)-3,4-dihydro-4-oxo-2-thioxo-1(2H)-pyrimidineacetamide
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.