Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 06409577
GREENER_CHOICE

Catalog No. 611410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-141-0 10 mg
61-145-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-141-0 Supplier Tocris Bioscience™ Supplier No. 6114/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective allosteric activator of AMPK

PF 06409577 is a potent and selective allosteric activator of AMPK (EC50 values are 7 and >40000 nM for AMPKα1β1γ1 and AMPKα1β2γ1, respectively). Exhibits minimal off-target effects on a panel of receptors, ion channels, PDEs and kinases. Improves kidney function and reduces proteinuria in a preclinical model of diabetic nephropathy. Orally bioavailable.

Specifications

Chemical Name or Material PF 06409577
CAS 1467057-23-3
Synonym 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C19H16ClNO3
Formula Weight 341.79
Physical Form Solid
Color Gray
Product Type Bioactive Small Molecule
Purity 99%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.