Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PF 3644022
GREENER_CHOICESDP

Catalog No. 427910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-791-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 42-791-0 Supplier R&D Systems™ Supplier No. 4279/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent MK2 inhibitor

Potent, ATP-competitive inhibitor of mitogen-activated protein kinase (MAPK)-activated protein kinase-2 (MK2) (IC50 = 5.2 nM; Ki = 3 nM). Inhibits tumor necrosis factor α (TNFα) production in U937 monocytic cells and peripheral blood mononuclear cells (PBMCs) (IC50 = 160 nM); exhibits oral efficacy in acute and chronic inflammatory models.

Chemical Identifiers

CAS 1276121-88-0
Molecular Formula C21H18N4OS

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 374.46
Purity >98%
Molecular Weight (g/mol) 374.462
Chemical Name or Material PF 3644022
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.