Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 3758309 dihydrochloride
GREENER_CHOICESDP

Catalog No. 600510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-051-0 10 mg
60-055-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-051-0 Supplier Tocris Bioscience™ Supplier No. 6005/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PAK4 inhibitor; orally available

PF 3758309 dihydrochloride is a potent PAK4 inhibitor (IC50 = 1.3 nM; Kd = 2.7 nM). Inhibits proliferation and anchorage-independent growth of a panel of tumor cell lines in vitro. Suppresses growth of human HCT116 and A549 tumor cell xenografts in mice. Orally available.

Specifications

Chemical Name or Material PF 3758309 Dihydrochloride
Synonym N-[(1S)-2-(Dimethylamino)-1-phenylethyl]-4,6-dihydro-6,6-dimethyl-3-[(2-methylthieno[3,2-d]pyrimidin-4-yl)amino]pyrrolo[3,4-c]pyrazole-5(1H)-carboxamide dihydrochloride
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Molecular Formula C25H30N8OS.2HCl
Formula Weight 563.55
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.