Learn More
Tocris Bioscience™ PF 3758309 dihydrochloride

Description
Specifications
Specifications
| Chemical Name or Material | PF 3758309 Dihydrochloride |
| Synonym | N-[(1S)-2-(Dimethylamino)-1-phenylethyl]-4,6-dihydro-6,6-dimethyl-3-[(2-methylthieno[3,2-d]pyrimidin-4-yl)amino]pyrrolo[3,4-c]pyrazole-5(1H)-carboxamide dihydrochloride |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in water and to 100mM in DMSO |
| Molecular Formula | C25H30N8OS.2HCl |
| Formula Weight | 563.55 |
| Physical Form | Solid |
| Color | Off-white |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.