Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PF 4800567 hydrochloride
GREENER_CHOICE

Catalog No. 428110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-811-0 10 mg
42-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-811-0 Supplier Tocris Bioscience™ Supplier No. 4281/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective casein kinase 1ε inhibitor

Selective casein kinase 1ε inhibitor; displays 22-fold greater potency towards CK1ε than CK1δ (IC50 values are 32 and 711 nM for CK1ε and CK1δ respectively). ATP competitive. Displays minimal effect on the circadian clock.

Chemical Identifiers

CAS 1391052-28-0
Molecular Formula C17H19Cl2N5O2

Specifications

Quantity 10 mg
Formula Weight 396.27
Purity >99%
Molecular Weight (g/mol) 396.272
Chemical Name or Material PF 4800567 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.