Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PF1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081671UV
Description
PF1 Polyclonal antibody specifically detects PF1 in Human, Mouse, Chicken samples. It is validated for Western BlotSpecifications
PF1 | |
Polyclonal | |
Western Blot | |
FLJ34122, KIAA1523MGC131914, PF1, PHD factor 1, PHD finger protein 12, PHD zinc finger transcription factor | |
The immunogen recognized by this antibody maps to a region between residue 954 and 1004 of human PHD factor 1 using the numbering given in entry NP_001028733.1 (GeneID 57649). | |
0.1 mL | |
Primary | |
Human, Mouse, Chicken | |
Purified |
Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
57649 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction