Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PF4 Monoclonal Antibody (C4)
Mouse Monoclonal Antibody
$184.40 - $493.00
Specifications
Antigen | PF4 |
---|---|
Clone | C4 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541661
|
Invitrogen™
MA541661 |
200 μL |
Each for $493.00
|
|
|||||
PIMA543756
|
Invitrogen™
MA543756 |
20 μL |
Each for $184.40
|
|
|||||
Description
PF4 Monoclonal Antibody for Western Blot, IHC (P)
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).Specifications
PF4 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P02776 | |
5196 | |
Recombinant protein Platelet Factor 4. The antigen corresponds to amino acid range 31-101 of the target protein. | |
Antibody |
C4 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
Pf4 | |
chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
Pf4 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title