Learn More
Invitrogen™ PF4 Monoclonal Antibody (RTO)

Description
MA5-17639 targets Platelet Factor 4 in ELISA and WB applications and shows reactivity with Human samples. The MA5-17639 immunogen is human Factor IV purified from the supernatant of thrombin activated platelets.
Specifications
Specifications
| Antigen | PF4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | RTO |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Pf4 |
| Gene Accession No. | P02776 |
| Gene Alias | chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.