Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554628
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554628 Supplier Invitrogen™ Supplier No. PA554628
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALERVLRLPA VASKRYLTNK VDRSVGGLVA QQQCVGPLQT PLADVAVVAL SHEELIGAAT ALGEQPVKSL LDPKVAARLA VAEALTNLVF ALVTDLRDVK CSG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

Purines are necessary for many cellular processes, including DNA replication, transcription, and energy metabolism. Ten enzymatic steps are required to synthesize inosine monophosphate (IMP) in the de novo pathway of purine biosynthesis. The enzyme encoded by this gene catalyzes the fourth step of IMP biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFAS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PFAS
Gene Accession No. O15067
Gene Alias 4432409B16Rik; FGAM synthase; FGAM synthetase; FGAMS; FGAR amidotransferase; FGARAT; FGAR-AT; Formylglycinamide ribonucleotide amidotransferase; formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Gm18; Kiaa0361; PFAS; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase); pur4; PURL; Sofa
Gene Symbols PFAS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PFAS. Recombinant protein control fragment (Product #RP-93470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5198
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.