Learn More
Invitrogen™ PFAS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110099
Description
Immunogen sequence: MSPVLHFYVR PSGHEGAAPG HTRRKLQGKL PELQGVETEL CYNVNWTAEA LPSAEETKKL MWLFGCPLLL DDVARESWLL PGSNDLLLEV GPRLNFSTPT STNIVSVCRA TGLGPVDRVE TTRRYRLSFA HPPSAEVEAI ALATLHDRMT EQHFPHPIQS FSPESMPEPL NGPINILGEG RLALEKANQE LGLALDSWDL DFYTKRFQEL QRNPSTVEAF DLAQSNSEHS RHWFFKGQLH VDGQKLVHSL FESIMSTQES SNPNNVLKFC DNSSAIQGKE VRFLRPEDPT RPSRFQQQQG.
Purines are necessary for many cellular processes, including DNA replication, transcription, and energy metabolism. Ten enzymatic steps are required to synthesize inosine monophosphate (IMP) in the de novo pathway of purine biosynthesis. The enzyme encoded by this gene catalyzes the fourth step of IMP biosynthesis.
Specifications
| PFAS | |
| Polyclonal | |
| Unconjugated | |
| PFAS | |
| 4432409B16Rik; FGAM synthase; FGAM synthetase; FGAMS; FGAR amidotransferase; FGARAT; FGAR-AT; Formylglycinamide ribonucleotide amidotransferase; formylglycinamide ribotide amidotransferase; formylglycinamide ribotide synthetase; Gm18; Kiaa0361; PFAS; Phosphoribosylformylglycinamidine synthase; phosphoribosylformylglycinamidine synthase (FGAR amidotransferase); pur4; PURL; Sofa | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 237823, 287420, 5198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15067, Q5SUR0 | |
| PFAS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PFAS (NP_036525.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.