Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN1 Antibody (12), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306609C
Description
PFDN1 Monoclonal antibody specifically detects PFDN1 in Human samples. It is validated for Western Blot, ELISASpecifications
PFDN1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
12 | |
Western Blot, ELISA | |
PDF, PFD1prefoldin 1, prefoldin subunit 1 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN1 (NP_002613.2; Met1-Gln122). | |
0.1 mL | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
5201 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction