Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297905FR
Description
PFDN1 Polyclonal specifically detects PFDN1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PFDN1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
PDF, PFD1prefoldin 1, prefoldin subunit 1 | |
Produced in rabbits immunized with E. coli-derived Human PFDN1 fragment. | |
0.1 mL | |
Cell Cycle and Replication, Membrane Trafficking and Chaperones | |
5201 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction