Learn More
Invitrogen™ PFDN1 Monoclonal Antibody (12)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PFDN1/Prefoldin subunit 1.
Specifications
Specifications
| Antigen | PFDN1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | PFDN1 |
| Gene Accession No. | O60925 |
| Gene Alias | 2700086I23Rik; AA408327; AU044714; PDF; PFD1; PFDN1; prefoldin 1; prefoldin subunit 1; Unknown (protein for MGC:128261) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.