Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552304 Supplier Invitrogen™ Supplier No. PA552304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELQAKVIDTQ QKVKLADIQI EQLNRTKKHA HLTDTEIMTL VDETNMYEGV GRMFILQSKE AIHSQLLEKQ KIAEEKIKEL EQKKSYLERS VKEAEDNIRE MLMARRA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PFDN1
Gene Accession No. O60925
Gene Alias 2700086I23Rik; AA408327; AU044714; PDF; PFD1; PFDN1; prefoldin 1; prefoldin subunit 1; Unknown (protein for MGC:128261)
Gene Symbols PFDN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PFDN1. Recombinant protein control fragment (Product #RP-89329).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.