Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN2 Antibody (04), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306549DL594
Description
PFDN2 Monoclonal antibody specifically detects PFDN2 in Human samples. It is validated for ImmunofluorescenceSpecifications
PFDN2 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
04 | |
Immunocytochemistry/Immunofluorescence | |
PFD2, prefoldin 2, prefoldin subunit 2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN2 (Q9UHV9; Met1-Ser154). | |
0.1 mL | |
Signal Transduction | |
5202 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction