Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300279MFV450
Description
PFDN2 Polyclonal antibody specifically detects PFDN2 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
PFDN2 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
PFD2, prefoldin 2, prefoldin subunit 2 | |
Produced in rabbits immunized with purified, recombinant Human PFDN2 (Uniprot#: Q9UHV9; Met1-Ser154) | |
0.1 mL | |
Signal Transduction | |
5202 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction