Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN2 Monoclonal Antibody (04), Invitrogen™

Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
| Antigen | PFDN2 |
|---|---|
| Clone | 04 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543485
|
Invitrogen™
MA543485 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543043
![]() |
Invitrogen™
MA543043 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human PFDN2
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.Specifications
| PFDN2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| ESTM27; HSPC231; PFD2; Pfdn2; prefoldin 2; prefoldin subunit 2; similar to prefoldin subunit 2; W48336 | |
| PFDN2 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 04 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UHV9 | |
| 5202 | |
| Recombinant Human PFDN2 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| PFDN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title