Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112647 Supplier Invitrogen™ Supplier No. PA5112647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PFDN2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PFDN2
Gene Accession No. Q9UHV9
Gene Alias ESTM27; HSPC231; PFD2; Pfdn2; prefoldin 2; prefoldin subunit 2; similar to prefoldin subunit 2; W48336
Gene Symbols PFDN2
Host Species Rabbit
Immunogen Recombinant Human Prefoldin subunit 2 protein (1-154AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5202
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.