Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31543
Description
Sequence: DVNVTFEDQQKINKFARNTSRITELKEEIEVKKKQLQNLEDACDDIMLADDDCLMIPYQIGDVFISHSQEETQEMLEEAKKNLQEEIDALESRVESIQRVLADLKVQLYAKFGSpecifications
PFDN4 | |
Polyclonal | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
prefoldin subunit 4 | |
PFDN4 | |
Recombinant protein corresponding to human PFDN4. | |
RUO | |
5203 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
C1/PFD4 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction