Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552583 Supplier Invitrogen™ Supplier No. PA552583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INITELNLPQ LEMLKNQLDQ EVEFLSTSIA QLKVVQTKYV EAKDCLNVLN KSNEGKELLV PLTSSMYVPG KLHDVEHVLI DVGTGYYVEK TAEDAKDFFK RKIDFLTKQM EKIQPALQEK HAMKQAVMEM MSQKIQQLTA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PFDN5
Gene Accession No. Q99471
Gene Alias 1190001O17Rik; 1700010A06Rik; c-myc binding protein; c-myc binding protein MM-1; C-myc-binding protein Mm-1; D15Ertd697e; Eig1; EIG-1; MM1; MM-1; Myc modulator 1; myc modulator-1; PFD5; Pfdn5; prefoldin 5; prefoldin subunit 5
Gene Symbols PFDN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PFDN5. Recombinant protein control fragment (Product #RP-89341).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5204
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.