Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592115 Supplier Invitrogen™ Supplier No. PA592115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQSINITEL NLPQLEMLKN QLDQEVEFLS TSIAQLKVVQ TKYVEAKDCL NVLNKSNEGK ELLVPLTSSM YVPGKLHDVE HVLIDVGTGY YVEKTAEDAK DFFKRKIDFL TKQMEKIQPA LQEKHAMKQA VMEMMSQKIQ QLTALGAAQA TAKA; Positive Samples: Raji, U-87MG, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PFDN5
Gene Accession No. Q99471, Q9WU28
Gene Alias 1190001O17Rik; 1700010A06Rik; c-myc binding protein; c-myc binding protein MM-1; C-myc-binding protein Mm-1; D15Ertd697e; Eig1; EIG-1; MM1; MM-1; Myc modulator 1; myc modulator-1; PFD5; Pfdn5; prefoldin 5; prefoldin subunit 5
Gene Symbols PFDN5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-154 of human PFDN5 (NP_0026152).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300257, 5204, 56612
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.