Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN5 Recombinant Rabbit Monoclonal Antibody (9Y5H7)
GREENER_CHOICE

Catalog No. PIMA542847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542847 Supplier Invitrogen™ Supplier No. MA542847
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: BxPC-3, RD, Jurkat, Mouse lung, Mouse testis, Mouse spleen, Rat lung, Rat testis, Rat spleen. Immunogen sequence: LNVLNKSNEG KELLVPLTSS MYVPGKLHDV EHVLIDVGTG YYVEKTAEDA KDFFKRKIDF LTKQMEKIQP ALQEKHAMKQ AVMEMMSQKI QQLTALGAAQ ATAKA.

Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9Y5H7
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PFDN5
Gene Accession No. Q99471, Q9WU28
Gene Alias 1190001O17Rik; 1700010A06Rik; c-myc binding protein; c-myc binding protein MM-1; C-myc-binding protein Mm-1; D15Ertd697e; Eig1; EIG-1; MM1; MM-1; Myc modulator 1; myc modulator-1; PFD5; Pfdn5; prefoldin 5; prefoldin subunit 5
Gene Symbols PFDN5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-154 of human PFDN5 (Q99471).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300257, 5204, 56612
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.