Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFDN6 Antibody (OTI4E10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273339B
Description
PFDN6 Monoclonal specifically detects PFDN6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
PFDN6 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
PFDN6 | |
Recombinant protein fragment corresponding to amino acids 2-129 of human PFDN6 (NP_055075) produced in E.coli. | |
0.1 mL | |
10471.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4E10 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
H2-KE2, HLA class II region expressed gene KE2, KE-2, KE2 protein, MGC70744, PFD6HKE2, prefoldin 6, prefoldin subunit 6, Protein Ke2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction