Learn More
PFDN6 Monoclonal Antibody (OTI4E10)

Description
PFDN6 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | PFDN6 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4E10 |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Pfdn6 |
| Gene Accession No. | O15212 |
| Gene Alias | H2-K region expressed gene 2; H-2Ke2; H2-KE2; HKE2; HLA class II region expressed gene KE2; hypothetical protein LOC393485; KE2; KE-2; KE2 protein; MHC class II region expressed gene KE2; Pfd6; PFDN6; prefoldin 6; prefoldin subunit 6; Protein Ke2; zgc:66282 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.