Learn More
PFDN6 Mouse anti-Human, Clone: OTI4E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504110

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. This protein also binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.Specifications
PFDN6 | |
Monoclonal | |
Unconjugated | |
PFDN6 | |
H2-KE2, HKE2, KE-2, PFD6 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10471 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15212 | |
PFDN6 | |
Human recombit protein fragment corresponding to amino acids 2-129 of human PFDN6 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.