Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PFDN6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561657
Description
Immunogen sequence: ARATVGKRLD YITAEIKRYE SQLRDLERQS EQQRETLAQL QQEFQRAQAA KAG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.
PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. This protein also binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
Specifications
PFDN6 | |
Polyclonal | |
Unconjugated | |
Pfdn6 | |
H2-K region expressed gene 2; H-2Ke2; H2-KE2; HKE2; HLA class II region expressed gene KE2; hypothetical protein LOC393485; KE2; KE-2; KE2 protein; MHC class II region expressed gene KE2; Pfd6; PFDN6; prefoldin 6; prefoldin subunit 6; Protein Ke2; zgc:66282 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10471 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15212 | |
Pfdn6 | |
Recombinant protein corresponding to Human PFDN6. Recombinant protein control fragment (Product #RP-99275). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction