Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFDN6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115741 Supplier Invitrogen™ Supplier No. PA5115741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PFD6.

PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. This protein also binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFDN6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Pfdn6
Gene Accession No. O15212, Q03958
Gene Alias H2-K region expressed gene 2; H-2Ke2; H2-KE2; HKE2; HLA class II region expressed gene KE2; hypothetical protein LOC393485; KE2; KE-2; KE2 protein; MHC class II region expressed gene KE2; Pfd6; PFDN6; prefoldin 6; prefoldin subunit 6; Protein Ke2; zgc:66282
Gene Symbols Pfdn6
Host Species Rabbit
Immunogen A synthesized peptide derived from human PFDN6(Accession O15212), corresponding to amino acid residues M1-L50.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10471, 14976, 309629
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.