Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PFI 1
GREENER_CHOICE

Catalog No. 444510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-451-0 10 mg
44-455-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-451-0 Supplier Tocris Bioscience™ Supplier No. 4445/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

BET bromodomain inhibitor

Potent BET bromodomain inhibitor; exhibits inhibitory activity at bromodomain-containing protein (BRD) 2 and BRD4 (IC50 values are 98 and 220 nM respectively). Induces apoptosis and G1 cell cycle arrest in BET inhibitor-sensitive cell lines (MV4;11). Also downregulates Aurora B expression in MV4;11 cells. Cell permeable.

Chemical Identifiers

CAS 1403764-72-6
Molecular Formula C16H17N3O4S

Specifications

Quantity 10 mg
Formula Weight 347.39
Purity >98%
Molecular Weight (g/mol) 347.389
Chemical Name or Material PFI 1
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.