Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFKFB2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PFKFB2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7337
![]() |
Bethyl Laboratories, Inc
A304286AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
PFKFB2 (6-phosphofructo-2-kinase/frustose-2, 6-biphosphatase 2) is responsible for the synthesis and degradation of fructose 2,6-biphosphate. Fructose 2, 6-biphosphate is a regulatory molecule that controls glycolysis in eukaryotes, mainly in the heart. PFKBF2 regulates the levels of fructose 2,6 biphosphate in the heart.Specifications
PFKFB2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
6PF-2-K/Fru-2,6-P2ase 2, 6PF-2-K/Fru-2,6-P2ASE heart-type isozyme, 6-phosphofructo-2-kinase, Fructose-2,6-bisphosphatase, fructose-2,6-bisphosphatase, cardiac isozyme, PFK/FBPase 2, PFK-2/FBPase-2, PFKFB, cardiac | |
Between 455 and 505 | |
Primary | |
4° C | |
PFKFB2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O60825, P70265 | |
18640, 5208 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title