Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PFKFB2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567335
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7335 Supplier Bethyl Laboratories, Inc Supplier No. A304286A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

PFKFB2 (6-phosphofructo-2-kinase/frustose-2, 6-biphosphatase 2) is responsible for the synthesis and degradation of fructose 2,6-biphosphate. Fructose 2, 6-biphosphate is a regulatory molecule that controls glycolysis in eukaryotes, mainly in the heart. PFKBF2 regulates the levels of fructose 2,6 biphosphate in the heart.

Specifications

Antigen PFKFB2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PFKFB2
Gene Accession No. O60825, P70265
Gene Alias 6PF-2-K/Fru-2,6-P2ase 2, 6PF-2-K/Fru-2,6-P2ASE heart-type isozyme, 6-phosphofructo-2-kinase, Fructose-2,6-bisphosphatase, fructose-2,6-bisphosphatase, cardiac isozyme, PFK/FBPase 2, PFK-2/FBPase-2, PFKFB, cardiac
Gene Symbols Pfkfb2
Host Species Rabbit
Immunogen Between 455 and 505
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18640, 5208
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.