Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PFKFB3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA515456
Description
This antibody is predicted to react with rat based on sequence homology.
The protein encoded by this gene belongs to a family of bifunctional proteins that are involved in both the synthesis and degradation of fructose-2,6-bisphosphate, a regulatory molecule that controls glycolysis in eukaryotes. The encoded protein has a 6-phosphofructo-2-kinase activity that catalyzes the synthesis of fructose-2,6-bisphosphate (F2,6BP), and a fructose-2,6-biphosphatase activity that catalyzes the degradation of F2,6BP. This protein is required for cell cycle progression and prevention of apoptosis. It functions as a regulator of cyclin-dependent kinase 1, linking glucose metabolism to cell proliferation and survival in tumor cells. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PFKFB3 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q16875 | |
| PFKFB3 | |
| KLH conjugated synthetic peptide between 454-484 amino acids from the C-terminal region of human PFKFB3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PFKFB3 | |
| 6-biphosphatase 3; 6PF-2-K/Fru-2,6-P2ase 3; 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme; 6PF-2-K/Fru-2,6-P2ASE brain-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 3; E330010H22Rik; Fructose-2; Fructose-2,6-bisphosphatase; fructose-6-phosphate,2-kinase/fructose-2, 6-bisphosphatase; inducible 6-phosphofructo-2-kinase; inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase; IPFK2; iPFK-2; PFK/FBPase 3; PFK2; PFKFB3; RB2K; renal carcinoma antigen NY-REN-56; RP11-298K24.3; uPFK-2 | |
| Rabbit | |
| Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 5209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction