Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFKFB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528648 Supplier Invitrogen™ Supplier No. PA528648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, mouse liver, rat liver. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (85%), Chicken (86%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Synthesis and degradation of fructose 2,6-bisphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKFB4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PFKFB4
Gene Accession No. P25114, Q16877, Q6DTY7
Gene Alias 6-biphosphatase-4; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ASE testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; Fructose-2,6-bisphosphatase; PFK/FBPase 4; Pfkfb4
Gene Symbols PFKFB4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PFKFB4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270198, 5210, 54283
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.