Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFKFB4 Recombinant Rabbit Monoclonal Antibody (HL2204)
GREENER_CHOICE

Catalog No. FEMA550430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550430 Supplier Invitrogen™ Supplier No. MA550430
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PFKFB4 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Synthesis and degradation of fructose 2,6-bisphosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKFB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2204
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PFKFB4
Gene Accession No. Q16877
Gene Alias 6-biphosphatase-4; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ASE testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; Fructose-2,6-bisphosphatase; PFK/FBPase 4; Pfkfb4
Gene Symbols PFKFB4
Host Species Rabbit
Immunogen Recombinant fragment of human PFKFB4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5210
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.