Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFKL Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232212R
Description
PFKL Polyclonal antibody specifically detects PFKL in Human samples. It is validated for Western BlotSpecifications
PFKL | |
Polyclonal | |
Western Blot | |
6-phosphofructokinase, liver type, DKFZp686G1648, DKFZp686L2097, EC 2.7.1, EC 2.7.1.11, FLJ30173, FLJ40909, human liver-type 1-phosphofructokinase, EC 2.7.1.1110Phosphohexokinase, liver-type 1-phosphofructokinase, PFK-B, Phosphofructo-1-kinase isozyme B, Phosphofructokinase 1, phosphofructokinase, liver, phosphohexokinase | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Liver using the numbering given in entry NP_002617.3 (GeneID 5211). | |
0.1 mL | |
Lipid and Metabolism | |
5211 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction