Learn More
Invitrogen™ PFKL Polyclonal Antibody

Description
PFKL Polyclonal Antibody detects endogenous levels of PFKL protein.
Specifications
Specifications
| Antigen | PFKL |
| Applications | Immunohistochemistry, Western Blot, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PFKL |
| Gene Accession No. | P12382, P17858, P30835 |
| Gene Alias | 6-phosphofructokinase type B; 6-phosphofructokinase, liver type; AA407869; ATP-dependent 6-phosphofructokinase, liver type; ATP-dependent 6-phosphofructokinase, liver type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; EC 2.7.1.11; HAMAP-Rule:MF_03184}; K6PL; liver-type 1-phosphofructokinase; Pfkb; PFK-B; Pfkl; Pfk-l; Phosphofructo-1-kinase isozyme B; phosphofructokinase 1; phosphofructokinase, liver; phosphofructokinase, liver type; phosphofructokinase, liver, B-type; phosphohexokinase; phosphohexokinase {ECO:0000255 |
| Gene Symbols | PFKL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.