Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PFKM Recombinant Rabbit Monoclonal Antibody (4Z7X1)
GREENER_CHOICE

Catalog No. PIMA535387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535387 Supplier Invitrogen™ Supplier No. MA535387
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: FVHEGYQGLV DGGDHIKEAT WESVSMMLQL GGTVIGSARC KDFREREGRL RAAYNLVKRG ITNLCVIGGD GSLTGADTFR SEWSDLLSDL QKAGKITDEE A.

Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFKM
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Z7X1
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PFKM
Gene Accession No. P08237, P47857, P47858
Gene Alias 6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; AI131669; ATP-dependent 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; GSD7; HAMAP-Rule:MF_03184}; PFK; PFK1; PFK-1; Pfk4; Pfk-4; Pfka; PFK-A; PFKM; Pfk-m; PFKX; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-A; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); phosphohexokinase; phosphohexokinase {ECO:0000255; PPP1R122; protein phosphatase 1, regulatory subunit 122
Gene Symbols PFKM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Fructose 6 Phosphate Kinase (P08237).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18642, 5213, 65152
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.