Learn More
Invitrogen™ PFKM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522172
Description
Recommended positive controls: 293T, mouse brain, rat brain. Predicted reactivity: Mouse (96%), Rat (94%), Xenopus laevis (89%), Dog (97%), Pig (95%), Chicken (88%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
Specifications
PFKM | |
Polyclonal | |
Unconjugated | |
PFKM | |
6-phosphofructo-1-kinase; 6-phosphofructokinase type A; 6-phosphofructokinase, muscle type; AI131669; ATP-dependent 6-phosphofructokinase, muscle type; ATP-dependent 6-phosphofructokinase, muscle type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; GSD7; HAMAP-Rule:MF_03184}; PFK; PFK1; PFK-1; Pfk4; Pfk-4; Pfka; PFK-A; PFKM; Pfk-m; PFKX; Phosphofructo-1-kinase isozyme A; phosphofructokinase 1; phosphofructokinase 4; phosphofructokinase M variant 3/4; phosphofructokinase muscle; phosphofructokinase, muscle; phosphofructokinase, polypeptide X; phosphofructokinase-1 A isozyme; phosphofructokinase-A; phosphofructokinase-M; phosphofructokinase-M (connecting peptide); phosphohexokinase; phosphohexokinase {ECO:0000255; PPP1R122; protein phosphatase 1, regulatory subunit 122 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18642, 5213, 65152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.92 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300, 0.01% thimerosal; pH 7 | |
P08237, P47857, P47858 | |
PFKM | |
Recombinant fragment corresponding to a region within amino acids 1 and 253 of PFK (muscle) (Uniprot ID#P08237). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.